2,6-Di-O-palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAA0757-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:112.0 to 116.0 deg-C
- Specific rotation [a]20/D:+19.0 to +24.0 deg (C=0.8, EtOH)
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:652.95 g/mol
CAS Number:4218-81-9
Formula:C₃₈H₆₈O₈
MDL Number:MFCD00059738