Azelaic dihydrazide ≥95.0% (by titrimetric analysis)
TCIAA0746-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:182.0 to 187.0 deg-C
:
:216.29 g/mol
:223-806-6
:4080-95-9
:C₉H₂₀N₄O₂
:MFCD00051529