N-Acetyl-D-valine ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0678-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-11.0 to -14.0 deg (C=1, AcOH)
- Solubility in Acetic acid:almost transparency
Specification Test Results
About this item
:
:159.19 g/mol
EINECS:241-860-9
CAS Number:17916-88-0
Formula:C₇H₁₃NO₃
MDL Number:MFCD00026457