Order Entry
Italy
ContactUsLinkComponent
3-Allyloxypropionic acid ≥95.0% (by GC)
Irritant
3-Allyloxypropionic acid ≥95.0% (by GC)
Catalog # TCIAA0657-10G
Supplier:  TCI
CAS Number:  
Irritant
3-Allyloxypropionic acid ≥95.0% (by GC)
Catalog # TCIAA0657-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :130.14 g/mol
Boiling Pt:192°C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:22577-15-7
Formula:C₆H₁₀O₃
MDL Number:MFCD00039537