Order Entry
Italy
ContactUsLinkComponent
Sodium-4-amino-1,5-naphthalenedisulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-amino-1,5-naphthalenedisulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0342-25G
Supplier:  TCI
CAS Number:  
Sodium-4-amino-1,5-naphthalenedisulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0342-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :325.28 g/mol
CAS Number:85328-80-9
Formula:C₁₀H₉NO₆S₂
MDL Number:MFCD00021519