
N,N'-Bis(2-aminobenzal)ethylenediamine ≥97.0% (by total nitrogen basis)
Catalog # TCIAA0261-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:180.0 to 185.0 deg-C
- Solubility in hot Toluene:very faint turbidity
Specification Test Results
About this item
:
:266.35 g/mol
:GHS07
CAS Number:4408-47-3
Formula:C₁₆H₁₈N₄
MDL Number:MFCD00059854