Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Aminobenzaldehyde polymer
3-Aminobenzaldehyde polymer
3-Aminobenzaldehyde polymer
TCIAA0259-25G
:
TCI
:
3-Aminobenzaldehyde polymer
TCIAA0259-25G
:
TCI
:
Pk:
25 g
Seq:
3308
Appearance:
Light yellow to Yellow to Orange powder to crystal
Total Nitrogen:
12.5 to 13.5 %
:
:
29159-23-7
:
C₇H₇NO
:
MFCD00147530