Order Entry
Italy
ContactUsLinkComponent
6,9-Diamino-2-ethoxyacridine lactate monohydrate ≥98.0% (by HPLC, titration analysis)
Irritant
6,9-Diamino-2-ethoxyacridine lactate monohydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0136-25G
Supplier:  TCI
CAS Number:  
Irritant
6,9-Diamino-2-ethoxyacridine lactate monohydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0136-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in hot Water:
    almost transparency
  • Water:
    4.0 to 6.0 %

Specification Test Results

About this item

 :
 :361.39 g/mol
Merck Index:14,03716
 :GHS07
CAS Number:6402-23-9
Formula:C₁₈H₂₃N₃O₅
MDL Number:MFCD00149646