Order Entry
Italy
ContactUsLinkComponent
 
N-Acetyl-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
N-Acetyl-D-phenylalanine ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    165.0 to 169.0 deg-C
  • Specific rotation [a]20/D:
    -39.0 to -43.0 deg (C=5, MeOH)

 

Frequently Bought Together