Order Entry
Italy
ContactUsLinkComponent
 
1-Acetyl-3-methylurea ≥98.0% (by total nitrogen basis)
  TCIAA0101-25G
 :  TCI
 :  
1-Acetyl-3-methylurea ≥98.0% (by total nitrogen basis)
  TCIAA0101-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    176.0 to 181.0 deg-C
  • Solubility in hot Water:
    almost transparency

 

 

 :
 :116.12 g/mol
 :623-59-6
 :C₄H₈N₂O₂
 :MFCD00026167