
N-Acetyl-ε-caprolactam ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1888-91-1
:
Formula:C₈H₁₃NO₂
MDL Number:MFCD00003262
:155.20 g/mol
Boiling Pt:134°C
EINECS:217-565-6
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together