1-Cetylpyridinium chloride monohydrate
Catalog # EHERC11075000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
Synonyms: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
:
:358.01 g/mol
EINECS:204-593-9
UN:2811
:GHS06
:GHS09
Storage Temperature:Ambient
CAS Number:6004-24-6
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977
Melting Pt:77…83 °C
ADR:6.1 I
Merck Index:13,02041