Order Entry
Italy
ContactUsLinkComponent
 
Flammable
2,2-Diethoxypropane stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.20391.0025
Supplier:  Merck
CAS Number:  
Flammable
2,2-Diethoxypropane stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.20391.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    132.20 g/mol
  • Boiling Point:
    114 °C (1013 hPa)
  • Flash Point:
    8 °C
  • Density:
    0.83 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :132.2 g/mol
Boiling Pt:61 °C (1013 hPa)
Density:0.826 g/cm³ (18 °C)
Storage Temperature:Ambient
ADR:3,II
 :M-Clarity™ quality level = MQ200
EINECS:204-808-6
UN:1993
 :GHS02
CAS Number:126-84-1
Formula:C₇H₁₆O₂
MDL Number:MFCD00009224