L(+)-Lysine monohydrate
Nome del fornitore: PanReac AppliChem
Jump to:
About this item
Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate
- CAS Number:39665-12-8
- Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
- MDL Number:MFCD00151035
- Molecular Weight:164.2 g/mol
- Melting Pt:225 °C
- Storage Temperature:Refrigerator
- EINECS:200-294-2
2 Options Available Below
Product Details & Documents
Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate
- CAS Number:39665-12-8
- Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
- MDL Number:MFCD00151035
- Molecular Weight:164.2 g/mol
- Melting Pt:225 °C
- Storage Temperature:Refrigerator
- EINECS:200-294-2
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



