Order Entry
Italy
ContactUsLinkComponent
Triammonium dodecamolybdophosphate hydrate
Irritant
Triammonium dodecamolybdophosphate hydrate
Certificates
undefined
Irritant
Triammonium dodecamolybdophosphate hydrate

Specification Test Results

  • Appearance:
    Yellow Powder
  • Chloride (Cl):
    ≤500 ppm
  • Sulfate (SO4):
    ≤250 ppm
  • Calcium (Ca):
    ≤200 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Insoluble matter:
    ≤0.01 %

Specification Test Results

Related Information

Frequently Bought Together