Order Entry
Italy
Orders LinkContactUsLinkComponent
Indium(III) nitrate hydrate (≥29% In)

Indium(III) nitrate hydrate (≥29% In)

Glass bottle for solids, 25 g
Codice catalogo A18252.14
Oxidizing
Irritant
Nome del fornitore:   Thermo Fisher Scientific
Indium(III) nitrate hydrate (≥29% In)
Codice catalogo A18252.14
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  207398-97-8
Indium(III) nitrate hydrate (≥29% In)
Indium(III) nitrate hydrate (≥29% In)

Glass bottle for solids, 25 g

Numero CAS:  207398-97-8
Price
155,00 €
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 207398-97-8 Molecular Formula: In(NO3)3.xH2O Formula Weight: 300.83(anhy) Melting Point: 100° -H2O UN No.: 1477 MDL No.: MFCD00149742

Synonyms:  Indium trinitrate hydrate
  • CAS Number:207398-97-8
  • Formula:InN₃O₉·H₂O
  • MDL Number:MFCD00149742
  • Molecular Weight:318.85 g/mol
  • Melting Pt:100 °C (-H₂O)
  • Storage Temperature:Ambient
  • EINECS:237-393-5
  • UN:1477
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications