Order Entry
Italy
ContactUsLinkComponent
Gallium trinitrate hydrate 99.9998% (trace metals basis)
Irritant
Oxidizing
Gallium trinitrate hydrate 99.9998% (trace metals basis)
Certificates
undefined
Irritant
Oxidizing
Gallium trinitrate hydrate 99.9998% (trace metals basis)

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Total trace metal impurities:
    ≤0.0002 %
  • Gallium (Ga):
    18 to 22 %

Specification Test Results

Related Information

Frequently Bought Together