

Gallium trinitrate hydrate, Sigma-Aldrich®
Catalog # 1.12171.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:65 °C
- pH:2 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:273.75 g/mol
ADR:5.1,
:M-Clarity™ quality level = MQ200
EINECS:236-815-5
UN:1477
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:69365-72-6
Formula:GaN₃O₉·H₂O
MDL Number:MFCD00149723