


Boron trifluoride dihydrate (65% BF₃)
:13319-75-0
:
:BF₃·2H₂O
:MFCD00149901
:103.84 g/mol
:58…60 °C (1.5 mmHg)
:6 °C
:1.642 g/cm³
:231-569-5
:2851
:8,II
:GHS07
:GHS08
:GHS05
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Assay:65.0 to 66.0 %
- Identification:Confirmed
Frequently Bought Together