Order Entry
Italy
ContactUsLinkComponent
Magnesium hydrogen orthophosphate trihydrate 98%, extra pure
Magnesium hydrogen orthophosphate trihydrate 98%, extra pure
  ACRO264780010
 :  
undefined
Magnesium hydrogen orthophosphate trihydrate 98%, extra pure
  ACRO264780010
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Appearance:
    White Powder
  • Titration Complexometric:
    ≥97.5 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Arsenic (As):
    ≤1 ppm
  • Iron (Fe):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤20 ppm

 

 

 :
 :174.33 g/mol
 :2.13 g/cm³
 :Ambient
 :231-823-5
 :7782-75-4
 :HMgO₄P·3H₂O
 :MFCD00150390