

Disodium metasilicate nonahydrate 44-47.5% (total solids)
:13517-24-3
:
:Na₂SiO₃·9H₂O
:MFCD00149175
:284.2 g/mol
:72 °C
:2,62 g/cm³ (21 °C)
:Ambient
:229-912-9
:3253
:8,III
:GHS05
:GHS07
- Appearance:White Granular powder or granules (May have a yellow-pink hue)
- Assay:44 to 47.5 % total solids
- Ratio:1.02 to 1.06 molar ratio SiO2/Na2O
- GC:≥96.0 %
Frequently Bought Together