
Lithium bromide hydrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:85017-82-9
:13453-70-8
Formula:BrLi·xH₂O
MDL Number:MFCD00149763
:86.85 g/mol (anhy)
Merck Index:14,5526
Boiling Pt:1265 °C (1013 hPa)
Melting Pt:162…167 °C
Storage Temperature:Ambient
EINECS:231-439-8
:GHS07
Frequently Bought Together