Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®

Irritant
Supplier:   Merck
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®
Supplier:  Merck
Certificates
2',4',6'-Trihydroxyacetophenone monohydrate, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  2-Acetylphloroglucinol monohydrate, THAP monohydrate, Phloroacetophenone monohydrate
  • CAS Number:207-556-5
  • Formula:C₈H₈O₄·1H₂O
  • MDL Number:MFCD00149091
  • Molecular Weight:186.16 g/mol
  • Boiling Pt:394 °C (1013 hPa)
  • Melting Pt:216…218 °C
  •  :M-Clarity™ quality level = MQ100
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 74,60 €
    74,60 €
    1 * 1 g
       74,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    91928-1G
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 282,00 €
    282,00 €
    1 * 5 g
       282,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    91928-5G