β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:145224-94-8
:
Formula:C₆H₁₃NO₄S·1H₂O
MDL Number:MFCD00149409
:213.25 g/mol
Merck Index:13,05929
Melting Pt:> 300 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:224-632-3
Related Information
Certificates
Search by Lot NumberMERC1.06126.0025_TDS_EN ProductPage_106126 ProductPage_106126
Product specification
msds_106126
SDS
Frequently Bought Together