Order Entry
Italy
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate 99+% for biochemistry
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate 99+% for biochemistry
Certificates
undefined
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate 99+% for biochemistry
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    8.0 to 9.0 % (K.F.)
  • UV:
    (0.1 M in water) 1 cm cell
  • pH:
    2.5 to 4.0 (1 % in water at 25°C)
  • Titration with NaOH:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together