VWR® ARISTAR®, Tellurium Standard Solution, 10000 mg/L Te in 40% Hydrochloric Acid (from Te), Standard for ICP
Catalog # 456132P
Nome del fornitore: VWR Chemicals
Numero CAS:
Specifiche
- Concentration:10000 mg/L
- Matrix:40% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Tellurium
- Symbol:Te
Specifiche
Informazioni su questo articolo
Te in HCl 20%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.