Order Entry
Italy
ContactUsLinkComponent
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Catalog # RPN4301
Supplier:  Cytiva
undefined
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Catalog # RPN4301
Supplier:  Cytiva

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    Horseradish Peroxidase Complex, biotinylated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Rabbit
  • Storage temperature:
    2 to 8°C
  • Pk:
    0,5 ml

Specifications

About this item

For use in ELISA systems. For ELISA, it is recommended that the conjugate should be diluted in the range 1:1000 to 1:4000.

Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: