Order Entry
Italy
ContactUsLinkComponent
Hydriodic acid 67%, EMSURE® for analysis, Supelco®
Corrosive
Hydriodic acid 67%, EMSURE® for analysis, Supelco®
Catalog # 1.00345.0250
Supplier:  Merck
CAS Number:  
Corrosive
Hydriodic acid 67%, EMSURE® for analysis, Supelco®
Catalog # 1.00345.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    127 °C (1013 hPa)
  • Density:
    1.97 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

Specification Test Results

About this item

 :
Boiling Pt:127 °C (1013 hPa)
Density:1.7 g/cm³ (20 °C)
EINECS:233-109-9
ADR:8,II
 :M-Clarity™ quality level = MQ300
 :127.91 g/mol
UN:1787
 :GHS05
CAS Number:10034-85-2
Formula:HI
MDL Number:MFCD00011347