Order Entry
Italy
Orders LinkContactUsLinkComponent
Orcein

Orcein

Glass bottle, 50 mL
Codice catalogo KPLI1971-01-01
Nome del fornitore:   SERACARE LIFE SCIENCES
Orcein
Codice catalogo KPLI1971-01-01
Nome del fornitore:  SERACARE LIFE SCIENCES
Numero CAS:  1400-62-0
Orcein

Glass bottle, 50 mL

Numero CAS:  1400-62-0
Price
Inquire For Price
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Orcein (from synthetic), Natural Red 28
  • CAS Number:1400-62-0
  • MDL Number:MFCD00062310
  • EINECS:215-750-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.