Rifampicin
Glass bottle for solids, 300 mg
Catalog # PROOICRS0409
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
- CAS Number:13292-46-1
- Formula:C₄₃H₅₈N₄O₁₂
- MDL Number:MFCD00151389
- Molecular Weight:822.95 g/mol
- Merck Index:12,08382
- Melting Pt:183…188 °C
- EINECS:236-312-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:300 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



