Order Entry
Italy
ContactUsLinkComponent
 
 
Hydriodic acid 57% analytical reagent
  20345.267
 :  
undefined
 
Hydriodic acid 57% analytical reagent
  20345.267
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Heavy metals (as Pb):
    Max. 10 ppm
  • Non-volatile residue:
    Max. 0.5 %
  • Cl + Br (as Cl):
    Max. 0.03 %
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm

 

 

 :
 :127 °C (1013 hPa)
 :1.7 g/cm³ (20 °C)
 :1787
 :8,II
 :127.91 g/mol
 :233-109-9
 :GHS05
 :10034-85-2
 :HI
 :MFCD00011347