Order Entry
Italy
ContactUsLinkComponent
Biotin-20-UTP 10 mM in buffer (Tris-HCl, pH 7.5)
Biotin-20-UTP 10 mM in buffer (Tris-HCl, pH 7.5)
Catalog # 40034.
Supplier:  Biotium
CAS Number:  
Biotin-20-UTP 10 mM in buffer (Tris-HCl, pH 7.5)
Catalog # 40034.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    25 µl
  • Seq:
    1166

Specifications

About this item

 :
 :1036.26 g/mol
Storage Temperature:Freezer
Formula:C₃₃H₅₁N₆O₂₂P₃SLi₄