Order Entry
Italy
ContactUsLinkComponent
 
Quinine sulphate dihydrate, TECHNICAL
Catalog # 27250.120
Supplier:  VWR Chemicals
CAS Number:  
undefined
Quinine sulphate dihydrate, TECHNICAL
Catalog # 27250.120
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :782.96 g/mol
Merck Index:12,03822
Melting Pt:219 °C
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-359-2
CAS Number:6119-70-6
 :207671-44-1
Formula:C₄₀H₅₀N₄O₈S·2H₂O
MDL Number:MFCD00150790