Order Entry
Italy
Orders LinkContactUsLinkComponent
Eriochrome Black T, TECHNICAL

Eriochrome Black T, TECHNICAL

Glass bottle, 100 g
Catalog # 26002.183
Irritant
Environment
Supplier:   VWR Chemicals
Eriochrome Black T, TECHNICAL
Catalog # 26002.183
Supplier:  VWR Chemicals
CAS Number:  1787-61-7
Eriochrome Black T, TECHNICAL
Eriochrome Black T, TECHNICAL

Glass bottle, 100 g

CAS Number:  1787-61-7
Price
156,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T
  • CAS Number:1787-61-7
  • Formula:C₂₀H₁₂N₃NaO₇S
  • MDL Number:MFCD00003935
  • Molecular Weight:461.39 g/mol
  • Merck Index:13,03702
  • Storage Temperature:Ambient
  • EINECS:217-250-3
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.