Aluminium oxide activated basic for chromatography
Catalog # 21012.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Identification:Passes test
- Particle size ( 48 µm):Max. 20 %
- Particle size ( 200 µm):Max. 1.5 %
Specification Test Results
About this item
:
:101.96 g/mol
EINECS:215-691-6
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424