Order Entry
Italy
ContactUsLinkComponent
Dextran, M.W. ~ 150,000, powder
Dextran, M.W. ~ 150,000, powder
Supplier:  VWR Chemicals
Certificates
undefined
Dextran, M.W. ~ 150,000, powder
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    White/almost white powder
  • Intrinsic viscosity (37°C):
    0.32 - 0.37
  • Loss on drying:
    Max. 10.0 %
  • pH range:
    4.0 - 7.0
  • Solubility in water (1 %):
    Passes test
  • Ashes:
    Max. 2.0 %

Specification Test Results

Related Information

Frequently Bought Together