Order Entry
Italy
ContactUsLinkComponent
Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed
Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed
Catalog # 305735B
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed
Catalog # 305735B
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Viscosity (20°C; 4 %; water):
    5,5 - 6,6 mPa.s
  • Degree of hydrolysis:
    Min. 98 %
  • Ignition residue (SO4):
    Max. 1 %
  • Loss on drying (105°C):
    Max. 5 %

Specification Test Results

About this item

 :
CAS Number:9002-89-5
 :25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922