Order Entry
Italy
ContactUsLinkComponent
Polyvinyl alcohol, M.W. 115,000 ≥88%, hydrolysed
Polyvinyl alcohol, M.W. 115,000 ≥88%, hydrolysed
Catalog # 297914D
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polyvinyl alcohol, M.W. 115,000 ≥88%, hydrolysed
Catalog # 297914D
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    White granular powder
  • pH (4 %):
    4.5 - 6.5
  • Viscosity (20°C; 4 %; water):
    46 - 54 mPa.s
  • Degree of hydrolysis:
    86.5 - 89 %
  • Ignition residue:
    Max. 0.5 %
  • Loss on drying (60°C):
    Max. 5 %
  • Organic volatile impurities:
    Max. 1 %

Specification Test Results

About this item

 :
CAS Number:9002-89-5
 :25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922