Order Entry
Italy
ContactUsLinkComponent
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM in TE buffer (pH 7.5)
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM in TE buffer (pH 7.5)
Catalog # 40036.
Supplier:  Biotium
CAS Number:  
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM in TE buffer (pH 7.5)
Catalog # 40036.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    50 µl
  • Seq:
    1202

Specifications

About this item

 :
 :885.42 g/mol
Storage Temperature:Freezer
CAS Number:136632-30-9
Formula:C₂₈H₄₂N₇O₁₆P₃SLi₄