VWR®, High temperature viscosity standards
Supplier: VWR Chemicals
Certificates
About this item
High Temperature viscosity standards are ideal for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
- Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/ml) given at all temperatures
- Standard 500 ml pack size, in tamper-evident packaging
- Fully traceable to national standards
- Ensures full compliance to ASTM and IP test method protocols
Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Caution: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
Frequently Bought Together