Undecane, Standard Material for GC
Catalog # TCIAS0283-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:1120-21-4
- Description:Undecane, Purity (GC): min. 99.5 %
- Formula:H₃C(CH₂)₉CH₃
- Description:Undecane
- Flash point:65 °C (lit.)
- Boiling point:195 °C
- Appearance:Colorless clear liquid
Specifications
About this item
- Specific Gravity (20/20): 0,74