Methyl decanoate, standard material for GC
Catalog # TCIAS0308-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:110-42-9
- Description:Methyl Decanoate, Purity (GC): min. 99.5 %
- Environmentally Preferable:
- Formula:C₁₁H₂₂O₂
- Description:Methyl Decanoate
- Flash point:"107 °C (lit.)"
- Boiling point:229 °C (lit.)
- Appearance:Colorless clear liquid