Hexadecane, Standard Material for GC
TCIAS0288-5ML
: TCI
:
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:544-76-3
- Description:Hexadecane, Purity (GC): min. 99.5 %
- Formula:H₃C(CH₂)₁₄CH₃
- Description:Hexadecane
- Flash point:"136 °C (lit.)"
- Boiling point:287 °C (lit.)
- Appearance:White or Colorless powder to lump to clear liquid