Order Entry
Italy
ContactUsLinkComponent
Food Color Testing Solutions A Set (includes permitted 12 food colors in Japan /each 4mL)
Food Color Testing Solutions A Set (includes permitted 12 food colors in Japan /each 4mL)
  TCIAF0118-1SET
 :  TCI
 :  
Food Color Testing Solutions A Set (includes permitted 12 food colors in Japan /each 4mL)
  TCIAF0118-1SET
 :  TCI
 :  

 

  • Pk:
    1 SET
  • Physical form:
    Liquid

 

 

  • MDL: MFCD00197816