Anti-ORF3a Rabbit Polyclonal Antibody
BWRLNCP0018
- Antibody type:Primary
- Antigen symbol:ORF3a
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:2019-nCoV
- Gene ID:25393..26220
- Accession no.:QHD43417.1
- Storage buffer:Rabbit IgG, 1mg/ml in PBS with 0.02% sodium azide, pH7.2
- Storage temperature:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:ORF3a, synthetic
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
- Size:100 µg
- Pk:100 µG
ORF3a polyclonal antibody detects endogenous levels of ORF3a protein
- For research and diagnosis
Type: Primary
Antigen: ORF3a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 2019-nCoV