


Toluene, extra pure, Burdick & Jackson™
Catalog # HONC65533-173KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:173 kg
- Seq:4648
Specifications
Specification Test Results
- Assay (GC):Min. 99.8 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.03 %
- Benzene (GC):Max. 0.02 %
Specification Test Results
About this item
:
:92.14 g/mol
Boiling Pt:110.6 °C
Density:0.867 g/cm3
Flash Pt:4 °C
UN:1294
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512