Hydrogen ionophore II, Selectophore™ ionophore, Supelco®
Catalog # 95295-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:345.60 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:70268-36-9
Formula:C₂₄H₄₃N
MDL Number:MFCD00144460