Carbonate ionophore VII, Selectophore™, function tested ionophore, Supelco®
Catalog # 93206-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:1016.24 g/mol
CAS Number:222310-82-9
Formula:C₅₈H₇₉F₆NO₇