Order Entry
Italy
ContactUsLinkComponent
Chloride ionophore IV, Selectophore™ ionophore, Supelco®
Chloride ionophore IV, Selectophore™ ionophore, Supelco®
Catalog # 92332-25MG-F
Supplier:  Merck
CAS Number:  
undefined
Chloride ionophore IV, Selectophore™ ionophore, Supelco®
Catalog # 92332-25MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :582.91 g/mol
CAS Number:187404-67-7
Formula:C₃₃H₅₀N₄OS₂
MDL Number:MFCD06201021