Order Entry
Italy
ContactUsLinkComponent
Tridodecylmethylammonium chloride, Selectophore™ ionophore, Supelco®
  91661-1G
 :  Merck
 :  
undefined
Tridodecylmethylammonium chloride, Selectophore™ ionophore, Supelco®
  91661-1G
 :  Merck
 :  
  • Pk:
    1 g
  • Seq:
    3047
 :
 :572.47 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
 :7173-54-8
 :[CH₃(CH₂)₁₁]₃N(Cl)CH₃
 :MFCD00043303